Learn More
Microbiologics™ MicroScan™ Rapid Anaerobic Panel QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5084L
 View more versions of this product
Catalog No. 23-017-054
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Multiple
Culture purpose and QC application
Clostridium perfringens ATCC 13124, bacteroides fragilis ATCC 25285, Clostridium sordellii ATCC 9714, Finegoldia magna ATCC 29328
MicroScan™ rapid anaerobic panel QC set
Multiple
CE, online certificate of analysis
Lyophilized Pellet
Lyfo disk™
6 Pellets
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months