Microbiologics™ MicroScan™ Synergies Plus - Gram Negative Panel QC Set

Microorganism sets for culture purposes and QC applications

Manufacturer: Microbiologics™ 5165L

 View more versions of this product

Catalog No. 23-017-079

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    For Use With (Application) Culture purpose and QC application
    ATCC® Multiple
    Format Lyophilized Pellet
    Includes Enterobacter aerogenes ATCC 51697, Escherichia coli ATCC 25922, Pseudomonas aeruginosa ATCC 27853, Enterococcus faecalis ATCC 29212, Escherichia coli ATCC 35218, Klebsiella oxytoca ATCC 49131, Shewanella putrefaciens ATCC 49138, Enterobacter cloacae subspecies cloacae ATCC 49141, Proteus vulgaris ATCC 49132
    Media Type Lyfo disk™
    Product Type LYFO DISK
    Quantity 6/Pk.
    Shelf Life 6 to 24 Months
    Species Multiple
    Storage Requirements 2° to 8°C
    Certifications/Compliance CE, online certificate of analysis

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit