Learn More
Microbiologics™ MicroScan™ Synergies Plus - Gram Negative Panel QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5165P
 View more versions of this product
Catalog No. 23-003-471
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Multiple
Culture purpose and QC application
Enterobacter aerogenes ATCC 51697, Escherichia coli ATCC 25922, Pseudomonas aeruginosa ATCC 27853, Enterococcus faecalis ATCC 29212, Escherichia coli ATCC 35218, Klebsiella oxytoca ATCC 49131, Shewanella putrefaciens ATCC 49138, Enterobacter cloacae subspecies cloacae ATCC 49141, Proteus vulgaris ATCC 49132
KWIK-STIK
Multiple
CE, online certificate of analysis
Lyophilized Pellet
Kwik-stik™
2/Pk.
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months