Learn More
Microbiologics™ MicroSEQ™ Microbial Identification System Validation Panel QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5185L
 View more versions of this product
Catalog No. 23-017-093
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Multiple
Culture purpose and QC application
Bacillus subtilis subspecies subtilis ATCC 6051, Staphylococcus epidermidis ATCC 12228, Aspergillus brasiliensis ATCC 16404, Candida albicans ATCC 10231, Escherichia coli ATCC 8739, Clostridium sporogenes ATCC 3584, Micrococcus luteus ATCC 10240, Staphylococcus aureus subspecies aureus ATCC 29737
LYFO DISK
Multiple
CE, online certificate of analysis
Lyophilized Pellet
Lyfo disk™
6/Pk.
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months