Learn More
Microbiologics™ Novobiocin Differential Test QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5056L
 View more versions of this product
Catalog No. 23-017-039
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Multiple
Culture purpose and QC application
Staphylococcus epidermidis ATCC 12228, Staphylococcus saprophyticus ATCC 49453
Novobiocin differential test QC set
Multiple
CE, online certificate of analysis
Lyophilized Pellet
Lyfo disk™
6 Pellets
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months