Learn More
Microbiologics™ Optochin Differential / Bile Solubility Test QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5057L
 View more versions of this product
Catalog No. 23-017-041
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Multiple
Culture purpose and QC application
Streptococcus pneumoniae ATCC 6305, Streptococcus gallolyticus ATCC 49147
LYFO-DISK
Multiple
CE, online certificate of analysis
Lyophilized Pellet
Lyfo disk™
6/Pk.
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months