Learn More
Microbiologics™ Oxidase Test QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5058P
 View more versions of this product
Catalog No. 23-003-3646
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Multiple
Culture purpose and QC application
Escherichia coli ATCC 25922, Pseudomonas aeruginosa ATCC 27853
KWIK-STIK
Multiple
CE, online certificate of analysis
Lyophilized Pellet
Kwik-stik™
2/Pk.
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months