Learn More
Microbiologics™ RapID ANA Panel QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5065L
 View more versions of this product
Catalog No. 23-017-049
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Multiple
Culture purpose and QC application
Clostridium sordellii ATCC 9714, Parabacteroides distasonis ATCC 8503, bacteroides uniformis ATCC 8492
LYFO DISK
Multiple
CE, online certificate of analysis
Lyophilized Pellet
Lyfo disk™
6/Pk.
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months