Microbiologics™ Vitek™ 2 AST-GN QC Set with Klebsiella

Microorganism sets for culture purposes and QC applications

Manufacturer: Microbiologics™ 5189L

 View more versions of this product

Catalog No. 23-017-099

  Check Availability
Add to Cart

    Specifications

    Format Lyophilized Pellet
    Product Type LYFO DISK
    Certifications/Compliance CE, online certificate of analysis
    For Use With (Application) Culture purpose and QC application
    ATCC® Multiple
    Includes Escherichia coli ATCC 25922, Pseudomonas aeruginosa ATCC 27853, Escherichia coli ATCC 35218, Klebsiella pneumoniae subspecies pneumoniae ATCC 700603
    Media Type LYFO DISK™
    Quantity 6 Pellets
    Shelf Life 6 to 24 Months
    Species Multiple
    Storage Requirements 2° to 8°C

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit