Learn More
Microbiologics™ Vitek 2 AST-GP QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5187K
 View more versions of this product
Catalog No. 23-017-094
  Check Availability
Add to cart
Disclaimers:
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications Description & Specifications
Specifications
Multiple
Culture purpose and QC application
Enterococcus faecalis ATCC 29212, Enterococcus faecalis ATCC 51299, Escherichia coli ATCC 35218, Staphylococcus aureus subspecies aureus ATCC BAA-976, Staphylococcus aureus subspecies aureus ATCC BAA-977, Staphylococcus aureus ATCC BAA-1026, Staphylococcus aureus subspecies aureus ATCC 29213, Streptococcus pneumoniae ATCC 49619
Vitek™ 2 AST-GP QC set
Multiple
CE, online certificate of analysis
Lyophilized Pellet
Kwik-stik™
6U
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months