Learn More
Microbiologics™ Vitek™ 2 CBC QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5209K
 View more versions of this product
Catalog No. 23-017-126
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Multiple
Culture purpose and QC application
Ochrobactrum anthropi ATCC BAA-749, Microbacterium testaceum ATCC 15829, Klebsiella oxytoca ATCC 700324, Arcanobacterium haemolyticum ATCC BAA-1784, Corynebacterium urealyticum ATCC 43044, Corynebacterium renale ATCC BAA-1785, Microbacterium paraoxydans ATCC BAA-1818, Microbacterium liquefaciens ATCC BAA-1819, Curtobacterium pusillum ATCC 19096, Cellulosimicrobium cellulans ATCC BAA-1816, Cellulosimicrobium cellulans ATCC BAA-1817
KWIK-STIK
Multiple
CE, online certificate of analysis
Lyophilized Pellet
Kwik-stik™
6/Pk.
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months