Microbiologics™ Vitek™ 2 CBC QC Set

Microorganism sets for culture purposes and QC applications

Manufacturer: Microbiologics™ 5209L

 View more versions of this product

Catalog No. 23-017-127

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Culture purpose and QC application
    ATCC® Multiple
    Format Lyophilized Pellet
    Includes Ochrobactrum anthropi ATCC BAA-749, Microbacterium testaceum ATCC 15829, Klebsiella oxytoca ATCC 700324, Arcanobacterium haemolyticum ATCC BAA-1784, Corynebacterium urealyticum ATCC 43044, Corynebacterium renale ATCC BAA-1785, Microbacterium paraoxydans ATCC BAA-1818, Microbacterium liquefaciens ATCC BAA-1819, Curtobacterium pusillum ATCC 19096, Cellulosimicrobium cellulans ATCC BAA-1816, Cellulosimicrobium cellulans ATCC BAA-1817
    Media Type Lyfo disk™
    Product Type LYFO DISK
    Quantity 6/Pk.
    Shelf Life 6 to 24 Months
    Species Multiple
    Storage Requirements 2° to 8°C
    Certifications/Compliance CE, online certificate of analysis

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit