Learn More
Microbiologics™ Vitek™ 2 Colorimetric GN QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5152K
 View more versions of this product
Catalog No. 23-017-076
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Multiple
Culture purpose and QC application
Acinetobacter baumannii ATCC BAA-747, Ochrobactrum anthropi ATCC BAA-749, Shigella sonnei ATCC 25931, Proteus vulgaris ATCC 6380, Enterobacter hormaechei ATCC 700323, Klebsiella oxytoca ATCC 700324, Stenotrophomonas maltophilia ATCC 17666
Vitek™ 2 colorimetric GN QC set
Multiple
CE, online certificate of analysis
Lyophilized Pellet
Kwik-stik™
6U
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months