Microbiologics™ Vitek™ 2 Colorimetric GP QC Set

Microorganism sets for culture purposes and QC applications

Manufacturer: Microbiologics™ 5151P

 View more versions of this product

Catalog No. 23-004-509

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    For Use With (Application) Culture purpose and QC application
    ATCC® Multiple
    Format Lyophilized Pellet
    Includes Streptococcus equi subspecies zooepidemicus ATCC 43079, Kocuria kristinae ATCC BAA-752, Listeria monocytogenes ATCC BAA-751, Staphylococcus saprophyticus ATCC BAA-750, Streptococcus thermophilus ATCC 19258, Staphylococcus aureus subspecies aureus ATCC 29213, Enterococcus casseliflavus ATCC 700327, Staphylococcus sciuri subspecies sciuri ATCC 29061
    Media Type Kwik-stik™
    Product Type Vitek™ 2 colorimetric GP QC set
    Quantity 2U
    Shelf Life 6 to 24 Months
    Species Multiple
    Storage Requirements 2° to 8°C
    Certifications/Compliance CE, online certificate of analysis

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit