Microbiologics™ Vitek™ 2 GP Comprehensive QC Set

Microorganism sets for culture purposes and QC applications

Manufacturer: Microbiologics™ 5193L

 View more versions of this product

Catalog No. 23-017-114

  Check Availability
Add to Cart

    Specifications

    Format Lyophilized Pellet
    Includes Streptococcus equi subspecies zooepidemicus ATCC 43079, Kocuria kristinae ATCC BAA-752, Listeria monocytogenes ATCC BAA-751, Staphylococcus saprophyticus ATCC BAA-750, Streptococcus salivarius subspecies thermophilus ATCC 19258, Enterococcus saccharolyticus ATCC 43076, Enterococcus casseliflavus ATCC 700327, Staphylococcus sciuri subspecies sciuri ATCC 29061, Streptococcus pneumoniae ATCC 49619
    Media Type Lyfo disk™
    Product Type Vitek™ 2 GP comprehensive QC set
    Quantity 6 Pellets
    Storage Requirements 2° to 8°C
    Certifications/Compliance CE, online certificate of analysis
    For Use With (Application) Culture purpose and QC application
    Shelf Life 6 to 24 Months

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit