Microbiologics™ VITEK 2™ KWIK-STIK™ AST-GP67 and AST-GP75 QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer: Microbiologics™ 5220K
 View more versions of this product
Catalog No. 23-017-220
  Check Availability
Add to Cart
Includes: Enterococcus faecalis ATCC™ 29212™, Enterococcus faecalis ATCC™ 51299™, Staphylococcus aureus subsp. aureus ATCC™ 29213™, Staphylococcus aureus subsp. aureus ATCC™ BAA-976™, Staphylococcus aureus subsp. aureus ATCC™ BAA-977™, Staphylococcus aureus ATCC™ BAA-1026™
    Specifications
    Specimen Type Multiple
    Quantity 6/Pk.
    Product Type Kwik-stik
    Includes Enterococcus faecalis ATCC™ 29212™, Enterococcus faecalis ATCC™ 51299™, Staphylococcus aureus subsp. aureus ATCC™ 29213™, Staphylococcus aureus subsp. aureus ATCC™ BAA-976™, Staphylococcus aureus subsp. aureus ATCC™ BAA-977™, Staphylococcus aureus ATCC™ BAA-1026™
    Media Type Kwik-stik™
    • KWIK-STIK™
    • Gives accurate, repeatable results
    • Microorganism, hydrating fluid and inoculating swab all in one self-contained device
    • Each device features single microorganism strain in lyophilized pellet, reservoir of hydrating fluid and inoculating swab
    • For added convenience, peel-off identification label is included for easy documentation
    • Available in packs of 2 or 6
  • Description & Specifications
Related Products
Product Content Feedback