Microbiologics™ VITEK 2™ KWIK-STIK™ AST-ST01 QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer: Microbiologics™ 5221P
 View more versions of this product
Catalog No. 23-017-225
  Check Availability
Add to Cart
Includes: Streptococcus pneumoniae ATCC™ 49619™, Staphylococcus aureus subsp. aureus ATCC™ 29213™, Staphylococcus aureus subsp. aureus ATCC™ BAA-977™
    Specifications
    Quantity 2/Pk.
    Includes Streptococcus pneumoniae ATCC™ 49619™, Staphylococcus aureus subsp. aureus ATCC™ 29213™, Staphylococcus aureus subsp. aureus ATCC™ BAA-977™
    Media Type Kwik-stik™
    Product Type Kwik-stik
    Species Multiple
    • KWIK-STIK™
    • Gives accurate, repeatable results
    • Microorganism, hydrating fluid and inoculating swab all in one self-contained device
    • Each device features single microorganism strain in lyophilized pellet, reservoir of hydrating fluid and inoculating swab
    • For added convenience, peel-off identification label is included for easy documentation
    • Available in packs of 2 or 6
  • Description & Specifications
Related Products
Product Content Feedback