Learn More
Microbiologics™ VITEK 2™ KWIK-STIK™ AST-ST01 QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5221K
 View more versions of this product
Catalog No. 23-017-223
  Check Availability
Add to cart
Includes: Streptococcus pneumoniae ATCC™ 49619™, Staphylococcus aureus subsp. aureus ATCC™ 29213™, Staphylococcus aureus subsp. aureus ATCC™ BAA-977™
Specifications
Description
  • KWIK-STIK™
  • Gives accurate, repeatable results
  • Microorganism, hydrating fluid and inoculating swab all in one self-contained device
  • Each device features single microorganism strain in lyophilized pellet, reservoir of hydrating fluid and inoculating swab
  • For added convenience, peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
Description & Specifications
Specifications
Streptococcus pneumoniae ATCC™ 49619™, Staphylococcus aureus subsp. aureus ATCC™ 29213™, Staphylococcus aureus subsp. aureus ATCC™ BAA-977™
Kwik-stik
Multiple
Kwik-stik™
6/Pk.