Learn More
Microbiologics™ Vitek™ 2 NH Comprehensive QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5194L
 View more versions of this product
Catalog No. 23-017-116
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
CE, online certificate of analysis
Lyophilized Pellet
LYFO DISK
Multiple
Culture purpose and QC application
Aggregatibacter aphrophilus ATCC 33389, Haemophilus influenzae ATCC 9007, Eikenella corrodens ATCC BAA-1152, Paenibacillus polymyxa ATCC 7070, Enterobacter aerogenes ATCC 13048, Staphylococcus epidermidis ATCC 12228, Neisseria gonorrhoeae ATCC 19424, Neisseria lactamica ATCC 23970, Oligella urethralis ATCC 17960
6/Pk.
2° to 8°C
Safety and Handling
ShelfLife : 6 to 24 Months