Learn More
Microbiologics™ Vitek™ 2 NH QC Set
Microorganism sets for culture purposes and QC applications
Manufacturer:  Microbiologics™ 5166P
 View more versions of this product
Catalog No. 23-015-280
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Multiple
Culture purpose and QC application
Aggregatibacter aphrophilus ATCC 33389, Haemophilus influenzae ATCC 9007, Campylobacter jejuni subspecies jejuni ATCC BAA-1153, Eikenella corrodens ATCC BAA-1152, Neisseria gonorrhoeae ATCC 19424, Neisseria lactamica ATCC 23970, Oligella urethralis ATCC 17960
Vitek™ 2 NH QC set
2° to 8°C
CE, online certificate of analysis
Lyophilized Pellet
Kwik-stik™
2U
Safety and Handling
ShelfLife : 6 to 24 Months