Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell™ MICROCHEM™ by AlphaTec™ Lab Coat
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
- Protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
- Comfortable to wear featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress
- Optimized fit, improving wearer comfort and safety
- Low Linting
Specifications
Type | Lab Coat |
Size | X-Large |
Material | Microporous Polyethylene Laminate |
Color | White |
Closure Type | Zipper |
No. of Pockets | 2 |
Latex Free | Free of natural rubber latex |
Seam Type | Bound |
Certifications/Compliance | CE CATEGORY III (Type PB (6)-B, EN 14126) |
For Use With (Application) | Clean room cleaning and preparing; Production line support and maintenance; Maintenance of plant and machines; Veterinary Services |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction