Learn More
Ansell™ MICROCHEM™ by AlphaTec™ Single-Use Chemical Suit
Ansell™ MICROCHEM™ by AlphaTec™ Single-Use Chemical Suit provides excellent chemical protection from critical environments
Manufacturer:  Ansell™ NV15S9210708
 View more versions of this product
Catalog No. 19-500-820
Add to cart
Description
Description
  • Proven protection, filters 99.9% of particles >3 microns
  • Air and water vapour permeable (breathable) fabric helps reduce the risk of heat stress
  • Optimised body fit, improving wearer comfort and safety
  • Silicone free, critical in spray painting applications
  • Anti-static, tested according to EN 1149-5 and AATCC 76
  • Applications:
    • Body surface checking, marking and touching up
    • Painting or cleaning tools or robots
    • Unpacking of bulk goods
    • Loading and unloading of process equipment
    • Production line support and maintenance
    • Assembly and inspection of components
    • Asbestos related work
    • Handing of powders
Specifications
Specifications
Attached boots, Elastic
Safety
Hooded
Elastic, Finger loops
Unisex
Hooded
1/Bag, 25/Cs.
4X-Large
50 gsm
Anti-static SMS
EN1073-2:2002; AATCC76; Type 5 EN ISO 13982-1:2004 + A1:2010; Type 6 EN13034:2005 + A1:2009
Blue
Yes
3-panel
Single use chemical suit
Serged
Hooded and booted coverall