Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ MICROCHEM™ by AlphaTec™ Single-Use Chemical Suit

Catalog Number p-7162836
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Ansell™ MICROCHEM™ by AlphaTec™ Single-Use Chemical Suit provides excellent chemical protection from critical environments

  • Proven protection, filters 99.9% of particles >3 microns
  • Air and water vapour permeable (breathable) fabric helps reduce the risk of heat stress
  • Optimised body fit, improving wearer comfort and safety
  • Silicone free, critical in spray painting applications
  • Anti-static, tested according to EN 1149-5 and AATCC 76
  • Applications:
    • Body surface checking, marking and touching up
    • Painting or cleaning tools or robots
    • Unpacking of bulk goods
    • Loading and unloading of process equipment
    • Production line support and maintenance
    • Assembly and inspection of components
    • Asbestos related work
    • Handing of powders
Product Type Chemical-resistant Coveralls
Size Small
Gender Unisex
Material Anti-static SMS
Color Blue
Disposable Single-use
Ankle Style Attached Boots, Elastic
Cuff Style Elastic, Finger Loops
Neck Style Hooded
Hood Type 3-panel
Seam Type Serged
Resistance Anti-static, Particle
Collar Type Hooded
Certifications/Compliance EN1073-2:2002; AATCC76; Type 5 EN ISO 13982-1:2004 + A1:2010; Type 6 EN13034:2005 + A1:2009
Thickness 50 gsm
Quantity 1/Bag, 25/Cs.
Style Hooded and booted coverall
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit