Learn More
Abnova™ Micrococcal nuclease (S. aureus)
Used for Enzyme
Manufacturer:  Abnova™ P5291
Catalog No. 89-952-481
Add to cart
Description
Description
#REF!
Specifications
Specifications
Native protein chromatographically purified from Staphylococcus aureus (Strain Foggi)
Lyophilized
Micrococcal nuclease (S. aureus)
15000 Units
RUO
Micrococcal Nuclease Assay; Elucidation of nucleic acid and oligonucleotide sequence studies.
Staphylococcus
Enzyme Activity
16.8kDa
Native protein chromatographically purified from Staphylococcus aureus (Strain Foggi)
Store at 4°C.
MICROCOCCAL
≥6,000 units per mg protein. One Unit corresponds to a change in optical density of 1.0 at 260nm at 37°C, pH 8.0, using DNA as the substrate.
None