Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma Microfil™ with EZ-Pak™, Black Filter

Catalog No. p-200104860
Click to view available options
Pore Size:
0.45 μm
0.8 μm

Black mixed cellulose esters gridded filter.

Microfil™ with EZ-Pak™ is an easy to use, fast, and accurate system for routine microbiological analysis. For the ultimate convenience and efficiency, combine the EZ-Pak™ system with the Microfil™ filtration unit. The Microfil™ filtration system is a simple and reliable solution for routine microbiological testing. These ready-to-use 100 mL Microfil™ funnels that come with black gridded EZ-Pak™ filter system eliminate the need for washing and sterilizing after each test thus making the equipment handling easier and efficient to trap microorganisms.

The push-fit design of the Microfil™ funnels seals tightly to manifold supports without the need for clamps or O-rings. Unlike with conventional filter holders, incomplete sealing at the membrane/funnel interface is eliminated resulting in consistent recovery of organisms.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) EZ-Fit Filtration Manifold
Material (Membrane) MCE
Pore Size 0.8 μm
Sample Volume 100 mL
Sterility Sterile
Grid Color Black
Type Filtration System
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.