Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell MICROFLEX™ ApexPro™ LC100 AP12 Nitrile Gloves
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Tested against fentanyl, and gastric acid (vomit) to simulate hazardous overdose situations
- Gloves consist of a dual layer, bi-color design (white exterior, black interior)
- Delivers advanced barrier protection (AQL 0.65)
- True pH balanced interior coating for improved hand health
- Triple washed
Specifications
Certifications/Compliance | ASTM D6319, ASTM D6978, FDA21 CFR 177-2600, NFPA 1999:2018, ISO 12485, ISO 9001 |
Chemotherapeutic Rated | Yes |
Applications | Healthcare |
AQL | 0.65 |
Color | White, Black |
Disposable | Single-use |
Exterior Finish | Fully Textured |
Finger Thickness (English) | 8.3 mil |
Finger Thickness (Metric) | 0.21 mm |
Latex Free | Yes |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction