Learn More
Ansell MICROFLEX™ ApexPro™ LC100 AP12 Nitrile Gloves
Dual-layer nitrile exam gloves with extended cuff provides protection against opioids and other hazardous substances.
$30.88
Specifications
Certifications/Compliance ASTM D6319, ASTM D6978, FDA21 CFR 177-2600, NFPA 1999:2018, ISO 12485, ISO 9001
Thickness (English) 8.3 mil
Thickness (Metric) 0.21 mm
Applications Healthcare
AQL 0.65
View More Specs
Products 7
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-500-971
View Documents
Ansell
AP121X
X-Small
Pack of 100 for $30.88
Case of 10 PK for $308.80
Add to cart
19-500-972
View Documents
Ansell
AP121
Small
Pack of 100 for $30.88
Case of 10 PK for $308.80
Add to cart
19-500-973
View Documents
Ansell
AP122
Medium
Pack of 100 for $30.88
Case of 10 PK for $308.80
Add to cart
19-500-974
View Documents
Ansell
AP123
Large
Pack of 100 for $30.88
Case of 10 PK for $308.80
Add to cart
19-500-975
View Documents
Ansell
AP124
X-Large
Pack of 100 for $30.88
Case of 10 PK for $308.80
Add to cart
19-500-976
View Documents
Ansell
AP125
2X-Large
Pack of 100 for $30.88
Case of 10 PK for $308.80
Add to cart
19-500-977
View Documents
Ansell
AP126
3X-Large
Pack of 100 for $30.88
Case of 10 PK for $308.80
Add to cart
Description
Description
  • Tested against fentanyl, and gastric acid (vomit) to simulate hazardous overdose situations
  • Gloves consist of a dual layer, bi-color design (white exterior, black interior)
  • Delivers advanced barrier protection (AQL 0.65)
  • True pH balanced interior coating for improved hand health
  • Triple washed
Specifications
Specifications
ASTM D6319, ASTM D6978, FDA21 CFR 177-2600, NFPA 1999:2018, ISO 12485, ISO 9001
0.21 mm
0.65
11.8 in.
20 MPa
0.21 mm
0.15 mm
8.3 mil
Healthcare
Fully Textured
30 cm
8.3 mil
5.9 mil