Learn More
Ansell™ MICROFLEX™ ApexPro™ XP100 APXP12
Ansell™ MICROFLEX™ ApexPro™ XP100 APXP12 provides dual-layer protection against opioids and hazardous substances that makes hazardous materials more visible.
Manufacturer:  Ansell™ APXP121
 View more versions of this product
Catalog No. 19-500-939
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Tested against both fentanyl and gastric acid (vomit) to simulate hazardous, real world overdose situations
  • Black exterior and gray interior to better see potentially hazardous powders
  • Feature a dual layer, dual color design
  • Deliver advanced barrier protection (AQL 0.65)
  • Triple washed, and have a true pH balanced interior coating for improved hand health
Specifications
Specifications
Safety
Fully Textured
Nitrile
22 MPa
Palm: 5.9 mil; Finger: 8.3 mil
Black, Gray
300 mm
Yes
Yes
Yes
Small
100/Box, 10 Boxes/Cs
Palm: 0.15 mm; Finger: 0.21 mm
11.8 in.
ASTM D6319; FDA21 CFR 177-2600; ISO 13485; ISO 9001; NFPA 1999:2018