Microflex™ Black Dragon™ Zero Nitrile Exam Gloves

Manufacturer: Microflex Corporation BD1005NPF

 View more versions of this product

Catalog No. 19-045-910

  Check Availability
Add to Cart

    Specifications

    Certifications ASTM D 6319
    Glove Cuff Style Beaded
    Thickness 3.5mil
    Disposable Yes
    Hand Ambidextrous
    Grade Medical
    Length (English) 9.6 in.
    Length (Metric) 24.38cm
    Material Nitrile
    Powdered No
    Size 2X-Large
    Sterile No
    Tensile Strength 42
    Coating Material Textured fingertips
    Color Black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit