Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microflex™ Black Dragon™ Zero Nitrile Exam Gloves

Nitrile exam gloves with textured fingertips. If you are concerned about tactile sensitivity, consider Black Dragon™ powder-free gloves. These gloves not only mask stains and provide better framing, they are also easy to put on and wear comfortably.

Manufacturer:  Microflex™ BD1001NPF

 View more versions of this product

Catalog No. 19-045-906



Due to product restrictions, please to purchase or view availability for this product.


Description

Description

 

  • Black color masks stains and provides a backdrop for powders, solvents and other work elements
  • Textured fingertips provide a secure grip on tools
Specifications

Specifications

Safety
Yes
Ambidextrous
Nitrile
42
Powder-free
Non-sterile
24.38 cm
Textured fingertips
Beaded/Rolled
Medical
Small
3.5 mil
Black
9.6 in.
ASTM D 6319
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit