Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microflex™ CE4-313 Nitrile Gloves

Class 10 Ambidextrous Nitrile Gloves

Manufacturer:  Microflex Corporation CE4313M

 View more versions of this product

Catalog No. 19-166-1238



  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

ISO Class 4
Textured
Ambidextrous
Medium
9.5cm
Powder-free
12 in.
12 x 3.74 in. (30.48 x 9.5cm)
Beaded/Rolled
Nitrile
3.74 in.
6.3mil
White
30.48cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit