Learn More
Ansell MICROFLEX™ Derma Free™ DF-850 Vinyl Gloves
Catalog Number p-138873
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Powder free, vinyl exam gloves feature a smooth surface for natural tactile sensitivity and are suitable for a variety of applications.
  • Made of 100% synthetic vinyl, eliminating the potential for adverse reactions associated with natural rubber latex proteins
  • Beaded cuff for easier donning and doffing
  • Powder free, ambidextrous
  • Nonsterile, single use only
  • Length: 9.1 in. (23.1 cm)
  • Thickness: 3.1 mil
  • Clear color
Applications Industrial
Size Small