Learn More
Microflex™ Synetron™ Latex High-Risk Lab Gloves
Thick and long enough to provide excellent protection
Manufacturer:  Microflex Corporation SY911M
 View more versions of this product
Catalog No. 18-999-4395B
  Check Availability
Add to cart
Alert:
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Specifications Description & Specifications
Specifications
Textured
Beaded/Rolled
Exam
Medium
ISO9001, ASTM D 3578, NFPA 1999
White
11.4 in.
Yes
Ambidextrous
Latex
11.8mil
Powder-free
Non-sterile
29cm