Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell MICROFLEX™ UltraSense™ US-220 Nitrile Gloves
GREENER_CHOICE

Catalog Number 19167032D
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Powder free, comfortable nitrile examination gloves offer durability when worn for extended periods of time. Ideal for healthcare, life science and food processing applications.

  • High-quality nitrile offers reliable barrier protection against many hazards and infectious substances
  • Ultra soft nitrile provides the fit and feel of latex
  • Textured fingers provide secure grip in wet and dry environments
  • Non-sterile, ambidextrous, single-use
  • Length: 9.6 in. (24.4 cm)
  • Fingertip thickness: 4.3 mil. Palm thickness: 3.1 mil
  • Unique dark blue color
Certifications/Compliance ASTM D6319, FDA21 CFR 177-2600
Certifications/Compliance (Europe) EN 374:2003, EN 420:2003 + A1:2009
Chemotherapeutic Rated No
Applications Industrial, Healthcare
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 4.3 mil
Finger Thickness (Metric) 0.11 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 3.1 mil
Palm Thickness (Metric) 0.08 mm
Powdered Powder Free
Size Large
Thickness (English) 3.1 mil
Thickness (Metric) 0.07 mm
FDA 510 (k) Yes
Model US-220-L
Series MICROFLEX™ UltraSense™ US-220
Tensile Strength 30 MPa
Length (English) 9.6 in.
Length (Metric) 24.38 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit