Learn More
Contec™ MicroGenesis Mop
MicroGenesis Mop 100% synthetic, disposable microfiber mop pad effectively removes fine particles and large debris from floors, providing superior removal of visible contaminants. Designed to be used wet or dry for general facilities cleaning in ISO Class 8 environments and above. It is compatible with a wide range of disinfectants, including quats, peroxides, and bleach. These synthetic pads are designed to provide uniform surface coverage.
Manufacturer:  Contec™ MGMP0003
 View more versions of this product
Catalog No. 19-165-902
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Synthetic construction is compatible with common disinfectants such as quats, peroxides and bleach
  • New pad for each use simplifies quality control, inventory and space management, all without cross-contamination risk
  • Compatible with most existing hardware
Specifications
Specifications
8 in.
6 in
Yes
Microfiber
6 x 8 in.
Non-sterile
Hand Pad