Learn More
Microlife Mix Culture
An opportunity to delve into the diversity of pond microorganisms.
Supplier: Neo/Sci Corporation 802001
Includes: Comprehensive identification sheet featuring over 50 illustrated microlife forms
Description
Just add water to this unique, dried mix to bring your own pond micro-ecosystem to life. Specially formulated mixture contains a variety of microhabitat materials, including bacteria and nutrients to support microlife growth.
This mix allows students to:
- Observe bacteria and other single-celled organisms within 24 hours
- Observe larger microinvertebrates within a week
- Get a view of a wide variety of microlife forms, including bacteria, green and non-green protists, filamentous algae, rotifers, ostracods and other microinvertebrates
- Safely stores for years without special handing