Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 11mm AlumiTin Magnetic Crimp Top Autosampler Vial Seals

MicroLiter's 11mm AlumiTin crimp-top autosampler vial seals are available in multiple colors and septa types.

Supplier:  DWK Life Sciences 110020RT

 View more versions of this product

Catalog No. 15-111-492

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

FEP/Natural Rubber
Aluminum
FEP/Natural Rubber
Crimp Top
11 mm Autosampler Vials - MicroLiter
Red
No
100
11 mm
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit