Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

DWK Life Sciences MicroLiter 11mm AlumiTin Magnetic Crimp Top Autosampler Vial Seals

MicroLiter's 11mm AlumiTin crimp-top autosampler vial seals are available in multiple colors and septa types.

Manufacturer:  DWK Life Sciences 110022YT

 View more versions of this product

Catalog No. 15-111-516



Description

Description

Specifications

Specifications

Yellow
No
ETFE/Silicone
Aluminum
100
11 mm Autosampler Vials - MicroLiter
ETFE/Silicone
11 mm Alumitin Seal
Crimp Top
11 mm
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit