Learn More
DWK Life Sciences MicroLiter 11mm AlumiTin Magnetic Crimp Top Autosampler Vial Seals
MicroLiter's 11mm AlumiTin crimp-top autosampler vial seals are available in multiple colors and septa types.
$32.75 - $775.00
Specifications
Closure Type | Crimp Top |
---|---|
Closure Material | Aluminum |
For Use With (Equipment) | 11 mm Autosampler Vials - MicroLiter |
Pre-slit | No |
Product Line | 11 mm Alumitin Seal |
Catalog Number | Mfr. No. | Closure Color | Septum | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Closure Color | Septum | Quantity | Price | Quantity & Availability | |||
15-111-485
|
DWK Life Sciences
110020AT |
Silver | FEP/Natural Rubber | 100 |
Pack of 100 for $32.75
|
|
15-111-487
|
DWK Life Sciences
110020BT |
Blue | FEP/Natural Rubber | 100 |
Pack of 100 for $45.00
|
|
15-111-492
|
DWK Life Sciences
110020RT |
Red | FEP/Natural Rubber | 100 |
Pack of 100 for $44.75
|
|
15-111-494
|
DWK Life Sciences
110020YT |
Yellow | FEP/Natural Rubber | 100 |
Pack of 100 for $45.00
|
|
15-111-497
|
DWK Life Sciences
110021AT |
Silver | FEP/Butyl | 100 |
Pack of 100 for $32.75
|
|
15-111-499
|
DWK Life Sciences
110021BT |
Blue | FEP/Butyl | 100 |
Pack of 100 for $40.50
|
|
15-111-503
|
DWK Life Sciences
110021RT |
Red | FEP/Butyl | 100 |
Pack of 100 for $40.50
|
|
15-111-505
|
DWK Life Sciences
110021YT |
Yellow | FEP/Butyl | 100 |
Pack of 100 for $40.50
|
|
15-111-508
|
DWK Life Sciences
110022AT |
Silver | ETFE/Silicone | 100 |
Pack of 100 for $32.75
|
|
15-111-510
|
DWK Life Sciences
110022BT |
Blue | ETFE/Silicone | 100 |
Pack of 100 for $40.50
|
|
15-111-512
|
DWK Life Sciences
110022GT |
Green | ETFE/Silicone | 100 |
Pack of 100 for $40.50
|
|
15-111-514
|
DWK Life Sciences
110022RT |
Red | ETFE/Silicone | 100 |
Pack of 100 for $40.50
|
|
15-111-516
|
DWK Life Sciences
110022YT |
Yellow | ETFE/Silicone | 100 |
Pack of 100 for $40.50
|
|
15-111-519
|
DWK Life Sciences
110030AT |
Silver | PTFE/Silicone | 100 |
Pack of 100 for $41.75
|
|
15-111-520
|
DWK Life Sciences
110030ATB |
Silver | PTFE/Silicone | 2000 |
Pack of 2000 for $775.00
|
|
15-111-525
|
DWK Life Sciences
110040AT |
Silver | PTFE/Silicone/PTFE | 100 |
Pack of 100 for $58.00
|
|
15-111-528
|
DWK Life Sciences
110040RT |
Red | PTFE/Silicone/PTFE | 100 |
Pack of 100 for $58.50
|
|
15-111-489
|
DWK Life Sciences
110020GT |
Green | FEP/Natural Rubber | 100 | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
15-111-501
|
DWK Life Sciences
110021GT |
Green | FEP/Butyl | 100 | This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Specifications
Crimp Top | |
11 mm Autosampler Vials - MicroLiter | |
11 mm Alumitin Seal |
Aluminum | |
No | |
11 mm |