Learn More
DWK Life Sciences MicroLiter 11mm AlumiTin Seal Crimp Top Autosampler Vial, Cap, and Septa Unassembled Component Kits
MicroLiter vials with AlumniTin seals are cleaned and packaged in a certified Class 10000 clean room according to a designed process, developed to eliminate chromatographic errors associated with vials
Manufacturer:  DWK Life Sciences 111030AT
 View more versions of this product
Catalog No. 15-111-544
Add to cart
Description
Description
  • Unassembled kits
  • Multiple configurations available
  • Identical, clean, and dependable, and are certified to ensure optimal performance and reproducible results
  • Critical physical dimensions are fully tested, as well as chemical cleanliness testing to ensure that trace levels of residues are not present
Specifications
Specifications
Silver
Borosilicate Glass (Type 1)
2 mL
PTFE/Silicone
Vial Kit
Crimp Top
11 mm
12 mm
No
PTFE/Silicone
100
Aluminum
32 mm