Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 11mm Crimp Top Autosampler Vials

MicroLiter 11mm Crimp Top Autosampler Vials are cleaned and packaged in a certified Class 10,000 clean room according to a designed process, developed to eliminate chromatographic errors associated with vials. Vials and inserts meet strict tolerances.

Manufacturer:  DWK Life Sciences 111500S

 View more versions of this product

Catalog No. 15-111-578


Add to cart

Description

Description

For use with Microliter 11mm Crimp Top Autosampler Vial seals
Specifications

Specifications

11mm
12 mm
High Recovery
11 mm Crimp-Top Seals - MicroLiter
Yes
Silanized Crimp uLvial
Clear
Borosilicate Glass (Type I)
1.8 mL
32mm
100
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit