Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 20mm AlumiTin Large Opening Crimp Top Headspace Vial Seals

MicroLiter 20mm large opening crimp top seals are available in multiple colors and septa types.

Manufacturer:  DWK Life Sciences 200025AT

 View more versions of this product

Catalog No. 15-111-674


Add to cart

Description

Description

Specifications

Specifications

Green
20 mm
20 mm Headspace Vials - MicroLiter
20 mm Alumitin Crimp Seal
Butyl stopper
Aluminum
Crimp Top
No
100
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit