Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 20 mm Crimp Top Headspace Vial Septas and Stoppers
SDP

Catalog No. 15111677 Shop All DWK Life Sciences Products
Click to view available options
Color:
Black
Blue, Natural
Gray
White
White, Tan
Material:
Aluminum, Silicone
Butyl
Molded PTFE, Butyl
PTFE, Butyl
PTFE, Silicone
Type:
3-leg Stopper
Septa
Stopper
No. per Pack:
100
1000

Butyl stoppers can be used when PTFE is not required. Butyl/PTFE septa are an economical choice with low background peaks.

Molded PTFE/butyl septa offer the tightest seal of all PTFE items, as well as low background peaks. PTFE/silicone septa are excellent for volatiles. Aluminum/silicone septa are also excellent for volatiles, but not suitable for repeat injections.
TRUSTED_SUSTAINABILITY

Specifications

Color Gray
Material PTFE, Butyl
Pre-slit No
No. per Pack 100
Type Septa
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Seals - MicroLiter
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.