Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 20mm Magnetic Crimp Top Headspace Vial Seals
SDP

MicroLiter 20mm crimp top seals are available in multiple colors and septa types.

Supplier:  DWK Life Sciences 200040MS

 View more versions of this product

Catalog No. 15-111-700

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

Magnetic crimp seal
Specifications

Specifications

PTFE/Molded Butyl
Steel
Molded PTFE/Butyl
Crimp Cap
20 mm Headspace Vials - MicroLiter
Gold
No
100
20 mm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit