Learn More
DWK Life Sciences MicroLiter 76 Series 9mm Autosampler Vial, Cap and Septa Unassembled Component Kits with FEP/Butyl Septa
Unassembled component kits offer the convenience of one product number for the vial, cap, and septa. Components are packaged to work with the unique MicroLiter Dispenser.
$51.09 - $56.96
Specifications
Volume (Metric) | 2 mL |
---|---|
Closure Material | Polypropylene |
Closure Type | Screw Thread |
Septum | FEP/Butyl |
Pre-slit | No |
Catalog Number | Mfr. No. | Material | Closure Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Closure Color | Price | Quantity & Availability | ||||
15-111-794
|
DWK Life Sciences
761021B |
Borosilicate Glass (Type 1) | Blue |
Pack of 100 for $51.09
|
|
15-111-796
|
DWK Life Sciences
761021G |
Borosilicate Glass (Type 1) | Green |
Pack of 100 for $51.09
|
|
15-111-798
|
DWK Life Sciences
761021K |
Borosilicate Glass (Type 1) | Black |
Pack of 100 for $51.09
|
|
15-111-800
|
DWK Life Sciences
761021N |
Borosilicate Glass (Type 1) | Natural |
Pack of 100 for $51.09
|
|
15-111-802
|
DWK Life Sciences
761021R |
Borosilicate Glass (Type 1) | Red |
Pack of 100 for $51.09
|
|
15-111-804
|
DWK Life Sciences
761021Y |
Borosilicate Glass (Type 1) | Yellow |
Pack of 100 for $51.09
|
|
15-111-893
|
DWK Life Sciences
761221B |
Borosilicate Glass (Type 1) | Blue |
Pack of 100 for $52.66
|
|
15-111-895
|
DWK Life Sciences
761221G |
Borosilicate Glass (Type 1) | Green |
Pack of 100 for $52.66
|
|
15-111-897
|
DWK Life Sciences
761221K |
Borosilicate Glass (Type 1) | Black |
Pack of 100 for $52.66
|
|
15-111-899
|
DWK Life Sciences
761221N |
Borosilicate Glass (Type 1) | Natural |
Pack of 100 for $52.66
|
|
15-111-901
|
DWK Life Sciences
761221R |
Borosilicate Glass (Type 1) | Red |
Pack of 100 for $52.66
|
|
15-111-903
|
DWK Life Sciences
761221Y |
Borosilicate Glass (Type 1) | Yellow |
Pack of 100 for $52.66
|
|
15-111-990
|
DWK Life Sciences
762221B |
Borosilicate Glass (Type 1) | Blue |
Pack of 100 for $56.96
|
|
15-111-992
|
DWK Life Sciences
762221G |
Borosilicate Glass (Type 1) | Green |
Pack of 100 for $56.96
|
|
15-111-994
|
DWK Life Sciences
762221K |
Borosilicate Glass (Type 1) | Black |
Pack of 100 for $56.96
|
|
15-111-996
|
DWK Life Sciences
762221N |
Borosilicate Glass (Type 1) | Natural |
Pack of 100 for $56.96
|
|
15-111-998
|
DWK Life Sciences
762221R |
Borosilicate Glass (Type 1) | Red |
Pack of 100 for $56.96
|
|
15-112-000
|
DWK Life Sciences
762221Y |
Borosilicate Glass (Type 1) | Yellow |
Pack of 100 for $56.96
|
|
Description
Available in multiple colors and septa formats and with or without writing patch.Specifications
2 mL | |
Screw Thread | |
No | |
9 mm |
Polypropylene | |
FEP/Butyl | |
76 Series Vial Kit |