Learn More
DWK Life Sciences MicroLiter 76 Series™ 9 mm Amber Glass Autosampler Vial, Cap and Septa Unassembled Component Kits with PTFE/Silicone Septa
SDP
Unassembled component kits offer the convenience of one product number for the vial, cap, and septa. Components are packaged to work with the unique MicroLiter Dispenser.
Supplier:  DWK Life Sciences 762235R
 View more versions of this product
Catalog No. 15-112-035
Add to Cart
Description
Description
Available in multiple colors and septa formats and with or without writing patch.
Specifications
Specifications
Red
12 mm
Yes
PTFE, Silicone
MicroLiter
Screw Cap
9 mm
Amber
Borosilicate Glass (Type 1)
2 mL
100
Polypropylene
32 mm
Yes
Product Suggestions
Product Suggestions