Learn More
DWK Life Sciences MicroLiter 76 Series™ 9mm Autosampler Vial, Cap and Septa Unassembled Component Kits w/ PTFE/Silicone/PTFE Septa
Unassembled component kits offer the convenience of one product number for the vial, cap, and septa. Components are packaged to work with the unique MicroLiter Dispenser.
Manufacturer:  DWK Life Sciences 761240K
 View more versions of this product
Catalog No. 15-111-950
Add to cart
Description
Description
Available in multiple colors and septa formats and with or without writing patch.
Specifications
Specifications
Black
Borosilicate Glass (Type 1)
2 mL
PTFE/Silicone/PTFE
76 Series Vial Kit
Screw Thread
9 mm
12 mm
No
PTFE/Silicone/PTFE
100
Polypropylene
32 mm