Learn More
DWK Life Sciences MicroLiter 9 mm Screw Thread Autosampler Vial, Cap, and Septa Assembled Kits with FEP/Butyl Septa
SDP
Save time and money with vial, cap, and septa kits that arrive conveniently packaged and preassembled.
Supplier:  DWK Life Sciences 091021GA
 View more versions of this product
Catalog No. 15-111-163
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Available in multiple colors and septa formats and with or without writing patch.
Specifications
Specifications
Green
12 mm
No
FEP, Butyl
MicroLiter
Screw Cap
9 mm
Clear
Borosilicate Glass (Type 1)
2 mL
100
Polypropylene
32 mm
No
Recommendations
Recommendations