Learn More
DWK Life Sciences MicroLiter 9mm Screwthread Autosampler Vial, Cap, and Septa Assembled Kits with FEP/Natural Rubber Septa
Save time and money with vial, cap, and septa kits that arrive conveniently packaged and preassembled.
Manufacturer:  DWK Life Sciences 092220BA
 View more versions of this product
Catalog No. 15-111-387
Add to cart
Description
Description
Available in multiple colors and septa formats and with or without writing patch.
Specifications
Specifications
Blue
Borosilicate Glass (Type 1)
2 mL
FEP/Natural Rubber/FEP
Assembled Kit
Screw Thread
9 mm
12 mm
No
FEP/Natural Rubber
100
Polypropylene
32 mm