Learn More
DWK Life Sciences MicroLiter 9mm Screwthread Autosampler Vial, Cap, and Septa Assembled Kits with PTFE/Silicone Septa
Save time and money with vial, cap, and septa kits that arrive conveniently packaged and preassembled.
Manufacturer:  DWK Life Sciences 091235GA
 View more versions of this product
Catalog No. 15-111-301
Add to cart
Description
Description
Available in multiple colors and septa formats and with or without writing patch.
Specifications
Specifications
Green
Borosilicate Glass (Type 1)
2 mL
PTFE/Silicone
Assembled Kit
Screw Thread
9 mm
12 mm
Yes
Precut PTFE/Silicone
100
Polypropylene
32 mm